Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
srsx-7
Entrez Gene ID
190819
Gene Name
Serpentine Receptor, class SX
Gene Aliases
CELE_Y97E10B.4
Chromosome Location
Chr.V: 7925783 - 7926920 on Build WBcel235
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001368455.1 NP_001355388.1 2 - 3 484 103

View Full Product Details