Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
glr-5
Entrez Gene ID
179251
Gene Name
GLutamate Receptor family (AMPA)
Gene Aliases
CELE_ZC196.7
Chromosome Location
Chr.V: 8716495 - 8721036 on Build WBcel235
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_072843.5 NP_505244.1 8 - 9 2024 77

View Full Product Details