Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
sru-30
Entrez Gene ID
183646
Gene Name
Serpentine Receptor, class U
Gene Aliases
CELE_C50C10.3
Chromosome Location
Chr.V: 9813827 - 9815007 on Build WBcel235
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_073132.2 NP_505533.1 1 - 2 247 111

View Full Product Details