Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
cash-1
Entrez Gene ID
179445
Gene Name
protein-coding
Gene Aliases
CELE_K07C5.8
Chromosome Location
Chr.V: 10362261 - 10365964 on Build WBcel235
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_073263.5 NP_505664.2 4 - 5 600 133

View Full Product Details