Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
plc-2
Entrez Gene ID
180029
Gene Name
Phosphoinositide phospholipase C
Gene Aliases
CELE_Y75B12B.6
Chromosome Location
Chr.V: 15192569 - 15201496 on Build WBcel235
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_074351.5 NP_506752.1 10 - 11 2183 78

View Full Product Details