Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
oac-7
Entrez Gene ID
180096
Gene Name
NRF domain-containing protein
Gene Aliases
CELE_C31A11.7
Chromosome Location
Chr.V: 16309805 - 16314775 on Build WBcel235
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_074719.5 NP_507120.3 7 - 8 1784 98

View Full Product Details