Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
srw-73
Entrez Gene ID
188019
Gene Name
G_PROTEIN_RECEP_F1_2 domain-containing protein
Gene Aliases
CELE_T03E6.6
Chromosome Location
Chr.V: 16586742 - 16588528 on Build WBcel235
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_074796.2 NP_507197.2 1 - 2 342 80

View Full Product Details