Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
srg-44
Entrez Gene ID
185159
Gene Name
Serpentine receptor class gamma
Gene Aliases
CELE_F31E9.2
Chromosome Location
Chr.V: 17328797 - 17330731 on Build WBcel235
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_074986.3 NP_507387.2 4 - 5 827 67

View Full Product Details