Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
par-4
Entrez Gene ID
180185
Gene Name
protein-coding
Gene Aliases
CELE_Y59A8B.14
Chromosome Location
Chr.V: 18098676 - 18106868 on Build WBcel235
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001380857.1 NP_001366977.1 8 - 9 1277 93
NM_001380858.1 NP_001367285.1 8 - 9 1254 93
NM_001380859.1 NP_001367284.1 5 - 6 843 93
GenBank mRNA AF160189.1 - 8 - 9 1300 93

Gene Symbol
Y73B3A.20
Entrez Gene ID
3564958
Gene Name
Uncharacterized protein
Gene Aliases
CELE_Y73B3A.20
Chromosome Location
Chr.X: 47766 - 58675 on Build WBcel235
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_075679.7 NP_508080.2 7 - 8 819 93

View Full Product Details