Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
ugt-46
Entrez Gene ID
180404
Gene Name
protein-coding
Gene Aliases
CELE_B0310.5
Chromosome Location
Chr.X: 525928 - 530289 on Build WBcel235
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001361781.3 NP_001348781.1 10 - 11 2368 109
NM_075711.7 NP_508112.1 9 - 10 1552 109

View Full Product Details