Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
set-30
Entrez Gene ID
180772
Gene Name
MYND-type domain-containing protein
Gene Aliases
CELE_ZC8.3
Chromosome Location
Chr.X: 4999270 - 5003516 on Build WBcel235
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_076449.6 NP_508850.2 2 - 3 226 143

View Full Product Details