Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
dct-11
Entrez Gene ID
189237
Gene Name
DAF-16/FOXO Controlled, germline Tumor affecting
Gene Aliases
CELE_W06B11.3
Chromosome Location
Chr.X: 5838039 - 5839778 on Build WBcel235
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_076578.8 NP_508979.2 5 - 6 660 73

View Full Product Details