Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
tnt-3
Entrez Gene ID
181052
Gene Name
protein-coding
Gene Aliases
CELE_C14F5.3
Chromosome Location
Chr.X: 7937633 - 7945099 on Build WBcel235
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001373380.2 NP_001359846.1 10 - 11 994 78
NM_001373382.2 NP_001360657.1 9 - 10 3668 78

View Full Product Details