Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
vab-15
Entrez Gene ID
181197
Gene Name
Homeobox protein vab-15
Gene Aliases
CELE_R07B1.1
Chromosome Location
Chr.X: 9847275 - 9849805 on Build WBcel235
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_077247.8 NP_509648.1 1 - 2 226 76
GenBank mRNA AF286218.1 - 1 - 2 224 76

View Full Product Details