Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
srd-38
Entrez Gene ID
187579
Gene Name
Serpentine Receptor, class D (delta)
Gene Aliases
CELE_R04D3.12
Chromosome Location
Chr.X: 13291806 - 13292984 on Build WBcel235
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_077810.1 NP_510211.1 2 - 3 434 117

View Full Product Details