Assay Design
The assay probe spans an exon junction. This assay will detect gDNA.
Gene Transcripts
- Gene Symbol
- gfi-3
- Entrez Gene ID
-
181544
- Gene Name
- Protein gfi-3
- Gene Aliases
-
CELE_M163.4
- Chromosome Location
-
Chr.X: 14480572 - 14484702 on Build WBcel235
Interrogated Sequence |
Translated Protein |
Exon Boundary |
Assay Location |
Amplicon Length |
RefSeq |
NM_078008.7 |
NP_510409.1 |
11 - 12 |
1353 |
109 |
- Gene Symbol
- M163.8
- Entrez Gene ID
-
3565205
- Gene Name
- Uncharacterized protein
- Gene Aliases
-
CELE_M163.8
- Chromosome Location
-
Chr.X: 14480582 - 14485955 on Build WBcel235
View Full Product Details