Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
ALPI
Entrez Gene ID
403736
Gene Name
alkaline phosphatase, intestinal
Gene Aliases
ALPP
Chromosome Location
Chr.25: 44107418 - 44110947 on Build CanFam3.1
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq XM_534605.7 XP_534605.5 4 - 5 1074 89
GenBank mRNA AF250845.1 - 3 - 4 410 89

View Full Product Details