Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
CCKAR
Entrez Gene ID
488846
Gene Name
cholecystokinin A receptor
Gene Aliases
-
Chromosome Location
Chr.3: 84010700 - 84019066 on Build CanFam3.1
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001012646.2 NP_001012664.1 1 - 2 112 150
GenBank mRNA AY934643.1 - 1 - 2 112 150

View Full Product Details