Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
KRTCAP2
Entrez Gene ID
480131
Gene Name
keratinocyte associated protein 2
Gene Aliases
KCP-2, KCP2
Chromosome Location
Chr.7: 42354793 - 42357596 on Build CanFam3.1
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001160122.1 NP_001153594.1 3 - 4 268 62
XM_022420757.1 XP_022276465.1 3 - 4 340 62
GenBank mRNA DN270212.1 - 3 - 4 268 62

View Full Product Details