Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
CHCHD2
Entrez Gene ID
479702
Gene Name
coiled-coil-helix-coiled-coil-helix domain containing 2
Gene Aliases
-
Chromosome Location
Chr.6: 581276 - 585367 on Build CanFam3.1
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq XM_536830.4 XP_536830.2 1 - 1 599 161

View Full Product Details