Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
SNRPD1
Entrez Gene ID
480178
Gene Name
small nuclear ribonucleoprotein D1 polypeptide
Gene Aliases
-
Chromosome Location
Chr.7: 66407313 - 66421193 on Build CanFam3.1
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001287157.1 NP_001274086.1 3 - 4 401 86
GenBank mRNA CO657835.1 - 1 - 2 189 86
CO672980.1 - 1 - 2 195 86
DT536786.1 - 3 - 4 387 86

View Full Product Details