Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
SRA1
Entrez Gene ID
606944
Gene Name
steroid receptor RNA activator 1
Gene Aliases
-
Chromosome Location
Chr.2: 35745518 - 35752497 on Build CanFam3.1
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq XM_005617283.3 XP_005617340.1 - 586 65

View Full Product Details