Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
PLPP7
Entrez Gene ID
491289
Gene Name
phospholipid phosphatase 7 (inactive)
Gene Aliases
-
Chromosome Location
Chr.9: 52781110 - 52796763 on Build CanFam3.1
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq XM_548410.6 XP_548410.3 1 - 2 891 59

View Full Product Details