Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
PPP1R11
Entrez Gene ID
608919
Gene Name
protein phosphatase 1 regulatory inhibitor subunit 11
Gene Aliases
-
Chromosome Location
Chr.35: 26289430 - 26292281 on Build CanFam3.1
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq XM_844699.5 XP_849792.1 1 - 2 305 63

View Full Product Details