Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
RIPPLY1
Entrez Gene ID
610288
Gene Name
ripply transcriptional repressor 1
Gene Aliases
-
Chromosome Location
Chr.X: 80457529 - 80461281 on Build CanFam3.1
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq XM_847751.5 XP_852844.3 1 - 2 667 60

View Full Product Details