Assay Design

The assay primers and probes lie within a single exon. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
ANKRD50
Entrez Gene ID
483835
Gene Name
ankyrin repeat domain 50
Gene Aliases
-
Chromosome Location
Chr.19: 15876656 - 15907899 on Build CanFam3.1
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq XM_540955.6 XP_540955.2 - 2903 75

View Full Product Details