Assay Design

The assay primers and probes lie within a single exon. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
TMEM53
Entrez Gene ID
482522
Gene Name
transmembrane protein 53
Gene Aliases
-
Chromosome Location
Chr.15: 15640232 - 15661829 on Build CanFam3.1
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq XM_005629089.3 XP_005629146.1 3 - 3 533 182
XM_022428035.1 XP_022283743.1 3 - 3 384 182
XM_022428037.1 XP_022283745.1 3 - 3 383 182

View Full Product Details