Assay Design

The assay primers and probes lie within a single exon. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
OR3A2
Entrez Gene ID
491211
Gene Name
olfactory receptor family 3 subfamily A member 2
Gene Aliases
OR10A04
Chromosome Location
Chr.9: 47191669 - 47192628 on Build CanFam3.1
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq XM_014116502.1 XP_013971977.1 1 - 1 632 198

View Full Product Details