Assay Design

The assay primers and probes lie within a single exon. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
SF3B5
Entrez Gene ID
100856528
Gene Name
splicing factor 3b subunit 5
Gene Aliases
-
Chromosome Location
Chr.1: 35558249 - 35558998 on Build CanFam3.1
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq XM_003638775.4 XP_003638823.1 - 276 99

View Full Product Details