Assay Design

The assay primers and probes lie within a single exon. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
MB21D2
Entrez Gene ID
100684351
Gene Name
Mab-21 domain containing 2
Gene Aliases
-
Chromosome Location
Chr.34: 24412254 - 24519829 on Build CanFam3.1
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq XM_005639839.3 XP_005639896.1 3 - 3 984 101
XM_022414169.1 XP_022269877.1 2 - 2 930 101
XM_022414170.1 XP_022269878.1 2 - 2 873 101

View Full Product Details