The assay primers and probes lie within a single exon. This assay will detect gDNA.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001313870.1 | NP_001300799.1 | - | 923 | 154 |
XM_005624295.3 | XP_005624352.1 | - | 1007 | 154 | |
XM_022422606.1 | XP_022278314.1 | - | 1096 | 154 | |
XM_022422607.1 | XP_022278315.1 | - | 898 | 154 |