Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
SCARA5
Entrez Gene ID
486097
Gene Name
scavenger receptor class A member 5
Gene Aliases
-
Chromosome Location
Chr.25: 29591248 - 29714159 on Build CanFam3.1
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq XM_005635672.3 XP_005635729.1 2 - 3 511 113
XM_543223.6 XP_543223.2 2 - 3 510 113

View Full Product Details