Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
SPACA3
Entrez Gene ID
610702
Gene Name
sperm acrosome associated 3
Gene Aliases
SPRASA
Chromosome Location
Chr.9: 40096178 - 40103848 on Build CanFam3.1
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001197087.1 NP_001184016.1 2 - 3 367 77
XM_005624359.2 XP_005624416.1 2 - 3 439 77
XM_005624360.3 XP_005624417.1 2 - 3 498 77
XM_022422604.1 XP_022278312.1 2 - 3 310 77
XM_022422605.1 XP_022278313.1 2 - 3 473 77
GenBank mRNA CX990262.1 - 3 - 4 411 77
FJ396442.1 - 2 - 3 355 77

View Full Product Details