Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
NPPA
Entrez Gene ID
608289
Gene Name
natriuretic peptide A
Gene Aliases
ANF, ANP, CDD
Chromosome Location
Chr.2: 84406317 - 84408115 on Build CanFam3.1
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001313851.1 NP_001300780.1 1 - 2 292 82

View Full Product Details