Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
Tk1
Entrez Gene ID
100689285
Gene Name
thymidine kinase 1, soluble
Gene Aliases
-
Chromosome Location
Chr.Un|NW_003613705: 623235 - 634238 on Build CriGri-PICRH-1.0
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001244494.1 NP_001231423.1 3 - 4 212 87
XM_007633677.1 XP_007631867.1 2 - 3 338 87
GenBank mRNA JI888091.1 - 3 - 4 269 87
JP058227.1 - 3 - 4 269 87

View Full Product Details