Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
Dhrs9
Entrez Gene ID
100771741
Gene Name
dehydrogenase/reductase (SDR family) member 9
Gene Aliases
-
Chromosome Location
Chr.Un|NW_003613606: 581752 - 604844 on Build CriGri-PICRH-1.0
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq XM_003495965.2 XP_003496013.1 4 - 5 991 93

View Full Product Details