Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
Bambi
Entrez Gene ID
100771599
Gene Name
BMP and activin membrane-bound inhibitor
Gene Aliases
I79_014040
Chromosome Location
Chr.Un|NW_003614263: 145852 - 150180 on Build CriGri-PICRH-1.0
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq XM_003506606.2 XP_003506654.1 2 - 3 378 61

View Full Product Details