Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
Apln
Entrez Gene ID
103161730
Gene Name
apelin
Gene Aliases
-
Chromosome Location
Chr.Un|NW_003613712: 668422 - 677704 on Build CriGri-PICRH-1.0
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq XM_007636362.1 XP_007634552.1 5 - 6 857 96

View Full Product Details