Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
Rusc1
Entrez Gene ID
100754395
Gene Name
RUN and SH3 domain containing 1
Gene Aliases
-
Chromosome Location
Chr.Un|NW_003613854: 265591 - 274569 on Build CriGri-PICRH-1.0
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq XM_003501581.2 XP_003501629.1 8 - 9 2550 85
XM_007642232.1 XP_007640422.1 7 - 8 1424 85
XM_007642233.1 XP_007640423.1 7 - 8 1187 85
XM_007642234.1 XP_007640424.1 6 - 7 1071 85
XM_007642235.1 XP_007640425.1 7 - 8 1143 85
XM_007642236.1 XP_007640426.1 7 - 8 1098 85
GenBank mRNA JI877155.1 - 1 - 2 217 85
JP047302.1 - 1 - 2 217 85

View Full Product Details