Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
Ripply1
Entrez Gene ID
100762657
Gene Name
ripply transcriptional repressor 1
Gene Aliases
I79_017114
Chromosome Location
Chr.Un|NW_003614629: 428050 - 434742 on Build CriGri-PICRH-1.0
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq XM_003509031.1 XP_003509079.1 4 - 5 428 143

View Full Product Details