Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
Bloc1s1
Entrez Gene ID
100772750
Gene Name
biogenesis of lysosomal organelles complex-1, subunit 1
Gene Aliases
I79_013525
Chromosome Location
Chr.Un|NW_003614217: 939775 - 943525 on Build CriGri-PICRH-1.0
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq XM_003506209.2 XP_003506257.1 2 - 3 241 89
GenBank mRNA JI883083.1 - 2 - 3 186 89

View Full Product Details