Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
Sspn
Entrez Gene ID
100758385
Gene Name
sarcospan
Gene Aliases
-
Chromosome Location
Chr.Un|NW_003614467: 511062 - 543939 on Build CriGri-PICRH-1.0
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq XM_003507996.2 XP_003508044.1 1 - 2 268 76

View Full Product Details