Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
Gxylt2
Entrez Gene ID
100758512
Gene Name
glucoside xylosyltransferase 2
Gene Aliases
-
Chromosome Location
Chr.Un|NW_003613585: 1424530 - 1483032 on Build CriGri-PICRH-1.0
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq XM_007620451.1 XP_007618641.1 4 - 5 776 73

View Full Product Details