Assay Design

The assay primers and probes lie within a single exon. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
Oxgr1
Entrez Gene ID
100761985
Gene Name
oxoglutarate (alpha-ketoglutarate) receptor 1
Gene Aliases
I79_000389
Chromosome Location
Chr.Un|NW_003613586: 1456405 - 1457421 on Build CriGri-PICRH-1.0
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq XM_003495227.1 XP_003495275.1 1 - 1 921 71

View Full Product Details