Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
Txndc17
Entrez Gene ID
100755189
Gene Name
thioredoxin domain containing 17
Gene Aliases
-
Chromosome Location
Chr.Un|NW_003614234: 740278 - 742904 on Build CriGri-PICRH-1.0
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq XM_003506367.2 XP_003506415.2 2 - 3 314 78
GenBank mRNA JI871236.1 - 2 - 3 517 78
JP060946.1 - 2 - 3 517 78

View Full Product Details