Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
Cnga2
Entrez Gene ID
100771809
Gene Name
cyclic nucleotide gated channel alpha 2
Gene Aliases
-
Chromosome Location
Chr.Un|NW_003614816: 23191 - 29239 on Build CriGri-PICRH-1.0
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq XM_003510036.1 XP_003510084.1 3 - 4 403 63

View Full Product Details