Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
Chchd2
Entrez Gene ID
100754041
Gene Name
coiled-coil-helix-coiled-coil-helix domain containing 2
Gene Aliases
-
Chromosome Location
Chr.Un|NW_003614718: 17492 - 24282 on Build CriGri-PICRH-1.0
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq XM_003509481.2 XP_003509529.1 3 - 4 555 80
GenBank mRNA JI892172.1 - 3 - 4 536 80
JP065756.1 - 3 - 4 536 80

View Full Product Details