Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
Mapre3
Entrez Gene ID
100761531
Gene Name
microtubule-associated protein, RP/EB family, member 3
Gene Aliases
-
Chromosome Location
Chr.Un|NW_003613640: 2063266 - 2067641 on Build CriGri-PICRH-1.0
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq XM_003497032.2 XP_003497080.1 3 - 4 520 101

View Full Product Details