Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
Neu4
Entrez Gene ID
100774175
Gene Name
sialidase 4
Gene Aliases
-
Chromosome Location
Chr.Un|NW_003613726: 1429890 - 1432653 on Build CriGri-PICRH-1.0
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq XM_003499265.2 XP_003499313.1 2 - 3 465 72

View Full Product Details