Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
Ndufa4
Entrez Gene ID
100766611
Gene Name
NADH dehydrogenase (ubiquinone) 1 alpha subcomplex, 4, 9kDa
Gene Aliases
I79_013258
Chromosome Location
Chr.Un|NW_003614203: 83293 - 91355 on Build CriGri-PICRH-1.0
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq XM_003505971.2 XP_003506019.1 1 - 2 174 69
GenBank mRNA JI894452.1 - 1 - 2 168 69

View Full Product Details